Skip to Content
Merck
All Photos(1)

Key Documents

54349

Sigma-Aldrich

Gold nanoparticles

0.6-0.9% solid material basis, 0.01% HAuCl4 in toluene

Synonym(s):

Dodecylthiol functionalized gold nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

description

nanoparticles

Quality Level

Assay

0.6-0.9% solid material basis

form

dispersion
nanoparticles

concentration

0.01% HAuCl4 in toluene

particle size

3-6 nm

functional group

thiol

SMILES string

CCCCCCCCCCCCS[Au]

InChI

1S/C12H26S.Au/c1-2-3-4-5-6-7-8-9-10-11-12-13;/h13H,2-12H2,1H3;/q;+1/p-1

InChI key

QMOCMJWMJUUCRE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Useful for the self-assembly of gold monolayers for nanoelectronics.

Features and Benefits

Surface stabilized with tetraoctylammonium bromide

Caution

Air-stable

Analysis Note

weak absorption peak at 520 nm

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service