Skip to Content
Merck
All Photos(1)

Key Documents

PRI444787724

Princeton BioMolecular Research, Inc.

5-Amino-1-(4-methyl-quinolin-2-yl)-1H-pyrazole-4-carboxylic acid amide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 528.25
250 MG
CHF 668.74
500 MG
CHF 851.37
1 G
CHF 1’088.79

Vendor SKU: PBMR156948-100MG

CHF 528.25


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 528.25
250 MG
CHF 668.74
500 MG
CHF 851.37
1 G
CHF 1’088.79

About This Item

CAS Number:
Molecular Weight:
267.29
MDL number:

Vendor SKU: PBMR156948-100MG

CHF 528.25


Ships in 14 days - from Aldrich Partner.

Iupac Name

5-amino-1-(4-methyl-2-quinolinyl)-1H-pyrazole-4-carboxamide

Inchi Code

1S/C14H13N5O/c1-8-6-12(18-11-5-3-2-4-9(8)11)19-13(15)10(7-17-19)14(16)20/h2-7H,15H2,1H3,(H2,16,20)

InChI key

FBNHKGKKUPSPSM-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

N/A

Product Link

http://store.princetonbio.com/?product_id=PBMR156948

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service