Skip to Content
Merck
All Photos(2)

Documents

SCC275

Sigma-Aldrich

721.221 Human HLA Negative B-Lymphoblastoid Cell Line

Human

Synonym(s):

721.221 Cell Line, B-Lymphoblastoid Cells, Human HLA Negative Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.45

product name

721.221 Human HLA Negative B-Lymphoblastoid Cell Line,

biological source

human

packaging

vial of >1X10⁶ cells

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Application

  • 721.221 Human HLA negative B-lymphoblastoid cells are verified to be of human origin and negative for mouse, rat, Chinese hamster, Golden Syrian hamster, and non-human primate interspecies contamination, as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells tested negative for infectious diseases against a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells tested negative for mycoplasma.

The human leukocyte antigen (HLA) gene complex encodes the major histocompatibility complex (MHC) proteins, key regulators of the human immune system. MHC proteins carry antigenic determinants for cell-surface recognition by T cells, and allow for immune autorecognition.1 The diversity of antigens provides a challenge for parsing individual T-cell activators and specific immune responses, and the ability to manipulate specific determinants is a paramount factor for understanding autoimmune diseases that arise from MHC dysfunction.
The 721.221 human HLA-negative B cell lymphoblastoid cell line is a well-established model for immune activation and HLA expression. The absence of class I HLA expression in 721.221 cells allows for functional analysis of individual class I antigens via transformation with individual HLA genes.2 HLA genes are ectopically expressed at quantitatively normal levels in 721.21 cells.2 721.221 cells express the B-cell marker CD19 4 and several NK activator ligands including CD48, CD80, and CD86. 5 The unique characteristics of the 721.221 cell line make it a highly versatile system for immunology research.

Source
The 721.221 cell line was derived from 721 lymphoblastoid cell line exposed to gamma radiation, inducing null mutations in endogenous HLA-A, B, and C class I antigen genes.

References
1. Simpson E. Immunol Suppl. 1988; 1:27-30.
2. Shimizu Y, DeMars R. J Immunol. 1989; 142(9):3320-3328.
3. Lisovsky I, Isitman G, Bruneau J, Bernard NF. J Leukoc Biol 2015; 97(4):761-767.
4. DeBell KE, Simhadri VR, Mariano JL, Borrego F. BMC Immunol 2012; 13:23.
5. Tremblay-McLean A, Coenraads S, Kiani Z, Dupuy FP, Bernard NF. BMC Immunol 2019; 20(1):8

Features and Benefits

The unique characteristics of the 721.221 cell line make it a highly versatile system for immunology research.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service