Skip to Content
Merck
All Photos(1)

Key Documents

MABS1165

Sigma-Aldrich

Anti-HSD17B3 Antibody, clone 3E11.2

clone 3E11.2, from mouse

Synonym(s):

Testosterone 17-beta-dehydrogenase 3, 17-beta-HSD 3, 17-beta-hydroxysteroid dehydrogenase type 3, Short chain dehydrogenase/reductase family 12C member 2, Testicular 17-beta-hydroxysteroid dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E11.2, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... HSD17B3(3293)

General description

Testosterone 17-beta-dehydrogenase 3 (EC 1.1.1.64; UniProt P37058; also known as 17-beta-HSD 3, 17-beta-hydroxysteroid dehydrogenase type 3, Short chain dehydrogenase/reductase family 12C member 2, Testicular 17-beta-hydroxysteroid dehydrogenase) is encoded by the HSD17B3 (also known as EDH17B3, SDR12C2) gene (Gene ID 3293) in human. 17 -hydroxysteroid dehydrogenases (17 -HSDs) mediate the activation and deactivation of sex steroids involving redox reactions centered about the C17 area of the steroid backbone. 17 -HSD3 catalyzes the final step in the steroidogenesis of the potent androgen testosterone (T) by selectively reducing the C17 ketone of 4-dione, utilizing NADPH as a cofactor, in testis and prostate. Upregulated 17 -HSD3 mRNA level is found in cancerous prostate biopsies when compared to normal tissue, while 17 -HSD3 deficiency caused by HSD17B3 gene mutations results in 46,XY Disorders of Sex Development (46,XY DSD).

Specificity

Clone 3E11.2 targets an epitope within the N-terminal half of mouse HSD17B3 and the two human spliced isoforms reported by UniProt (P70385 and P37058).

Immunogen

KLH-conjugated linear peptide corresponding to an internal sequence within the N-terminal half of human HSD17B3.

Application

Detect HSD17B3 using this mouse monoclonal Anti-HSD17B3 Antibody, clone 3E11.2, Cat. No. MABS1165, validated for use in Western Blotting.
Research Category
Signaling

Quality

Evaluated by Western Blotting in mouse testis tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HSD17B3 in 10 µg of mouse testis tissue lysate.

Target description

~37 kDa observed. 34.52/29.11 kDa (human isoform 1/2) and 34.33 kDa (mouse) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service