Skip to Content
Merck
All Photos(1)

Documents

AG253

Sigma-Aldrich

Heme Oxygenase 1, control peptide for AB1284

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
eCl@ss:
32160405
NACRES:
NA.41

Assay

88% (HPLC)

Quality Level

technique(s)

inhibition assay: suitable

UniProt accession no.

Gene Information

General description

FUNCTION: SwissProt: P09601 # Heme oxygenase cleaves the heme ring at the alpha methene bridge to form biliverdin. Biliverdin is subsequently converted to bilirubin by biliverdin reductase. Under physiological conditions, the activity of heme oxygenase is highest in the spleen, where senescent erythrocytes are sequestrated and destroyed.
SIZE: 288 amino acids; 32819 Da
SUBCELLULAR LOCATION: Microsome.
SIMILARITY: SwissProt: P09601 ## Belongs to the heme oxygenase family.

Application

Antibody blocking (AB1284)

Optimal working dilution must be determined by the end user.

Physical form

White lyophilized powder. Soluble in water.

Storage and Stability

Long term storage at -20°C in a sealed container with desiccant.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service