Skip to Content
Merck
All Photos(2)

Key Documents

ABN1365

Sigma-Aldrich

Anti-OSTM1

from rabbit, purified by affinity chromatography

Synonym(s):

Osteopetrosis-associated transmembrane protein 1, Chloride channel 7 beta subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), baboon (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... OSTM1(28962)

General description

Osteopetrosis-associated transmembrane protein 1 (UniProt: Q86WC4; also known as Chloride channel 7 beta subunit) is encoded by the OSTM1 (also known as GL) gene (Gene ID: 28962) in human. Osteopetrosis-associated transmembrane protein 1 is a single-pass type I membrane protein that is required for osteoclast and melanocyte maturation and function. It is synthesized with a signal peptide of 31 amino acids, which is cleaved to produce the mature protein. Loss of OSTM1 leads to the most severe form of osteoporosis in mice and human. Although its role in bone chemistry is well established, several studies have shown its expression in neurons, microglia, and astrocytes. OSTM1-deficiecy in the central nervous system leads to gliosis with increased astrocyte cell population and microglia activation along with retinal degeneration at 2 to 3 weeks of age in mice. (Ref.: Ref.: Heraud, C., et al. 2014. J. Biol. Chem. 289(20); 13912-25).

Specificity

This rabbit polyclonal antibody detects Osteopetrosis-associated transmembrane protein 1 in human cells. It targets an epitope within 18 amino acids from the C-terminus region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminus region of human Osteopetrosis-associated transmembrane protein 1 (OSTM1).

Application

Anti-OSTM1, Cat. No. ABN1365, is a highly specific rabbit polyclonal antibody that targets Osteopetrosis-associated transmembrane protein 1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected OSTM1 in human pancreas and human liver tissues.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected OSTM1 in 20 µg of human liver tissue lysate.

Target description

~37 kDa observed; 37.26 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, pH 7.4 with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service