Skip to Content
Merck
All Photos(1)

Key Documents

650297

Sigma-Aldrich

Anti-Trypsin Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
CHF 526.00

CHF 526.00


Estimated to ship on23 April 2025


Request a Bulk Order

Select a Size

Change View
1 ML
CHF 526.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

CHF 526.00


Estimated to ship on23 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Rabbit polyclonal antibody was purified by ammonium sulfate precipitation followed by DEAE-cellulose ion exchange chromatography. Recognizes cationic trypsin.
Recognizes cationic trypsin in pancreas tissue. Does not cross-react with other plasma proteins.
This Anti-Trypsin Rabbit pAb is validated for use in ELISA, Immunoblotting, Immunoelectrophoresis for the detection of Trypsin.

Immunogen

Human
human pancreatic trypsin

Application

ELISA (≥1:250)

Immunoblotting (1:2000)

Immunoelectrophoresis (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Human pancreas tissue

Other Notes

Does not cross-react with other plasma proteins. Monospecific as determined by immunoelectrophoresis against human pancreatic extract and against human trypsin. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Hye Seo et al.
Digestion, 80(1), 40-49 (2009-05-30)
Proteinase-activated receptor-2 (PAR2), which is activated by trypsin, is known to be associated with expression of adhesion molecule integrins. We previously demonstrated that Helicobacter pylori induced the expression of integrin alpha(5) and beta(1) in human gastric epithelial cells. The present
Carlie S Sigel et al.
Cancer cytopathology, 121(8), 459-470 (2013-02-15)
Acinar cell neoplasms of the pancreas are rare but when encountered, the diagnosis is often established based on cytology specimens. Diagnostic accuracy is important because acinar cell carcinomas are aggressive yet may mimic tumors with different outcomes and management. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service