Skip to Content
Merck
All Photos(1)

Key Documents

648494

Sigma-Aldrich

TRPC3 Channel Inhibitor III, Pyr10

Synonym(s):

TRPC3 Channel Inhibitor III, Pyr10, N-(4-(3,5-bis(trifluoromethyl)-1H-pyrazol-1-yl)phenyl)-4-methylbenzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 184.00

CHF 184.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
CHF 184.00

About This Item

Empirical Formula (Hill Notation):
C18H13F6N3O2S
CAS Number:
Molecular Weight:
449.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 184.00


Estimated to ship on16 April 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

semisolid
solid

potency

720 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange-brown
white to off-white

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

FC(F)(F)c1[n](nc(c1)C(F)(F)F)c2ccc(cc2)N[S](=O)(=O)c3ccc(cc3)C

InChI

1S/C18H13F6N3O2S/c1-11-2-8-14(9-3-11)30(28,29)26-12-4-6-13(7-5-12)27-16(18(22,23)24)10-15(25-27)17(19,20)21/h2-10,26H,1H3

InChI key

CVALMMCIOLXHDM-UHFFFAOYSA-N

General description

A cell-permeable pyrazole derivative that acts as a selective blocker of the transient receptor potential cation channel TRPC3-medated ROCE (receptor-operated Ca2+ entry) and inhibits Carbachol- (Cat. No. 212385) induced Ca2+ entry in TRPC3-expressing HEK293 cells (IC50 = 0.72 µM; by Fura-2), while exhibiting much reduced potency against Stim1 and Orai1-coupled CRAC (Ca2+ release-activated Ca2+) channel-mediated SOCE (store-operated Ca2+ entry) in RBL-2H3 cells (IC50 = 13.08 µM; by Fura-2). A great complement to Pyr2 (Cat. No. 203890), Pyr 3 (Cat. No. 648490), and Pyr6 (Cat. No. 203891) in Ca2+ signaling studies.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TRPC3
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Schleifer, H. et al. 2012. Br. J. Pharmacol.167, 1712.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service