Skip to Content
Merck
All Photos(1)

Key Documents

5.04467

Sigma-Aldrich

L-CCG-I

Synonym(s):

L-CCG-I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO4
CAS Number:
Molecular Weight:
159.14
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

aqueous base: soluble

storage temp.

2-8°C

InChI

1S/C6H9NO4/c7-4(6(10)11)2-1-3(2)5(8)9/h2-4H,1,7H2,(H,8,9)(H,10,11)/t2-,3-,4-/m0/s1

InChI key

GZOVEPYOCJWRFC-HZLVTQRSSA-N

General description

A potent agonist for group II metabotropic glutamate receptors (EC50 = 0.49 µM and 0.44 µM for mGlu2 and mGlu3, respectively). Higher level of activity than glutamate or (±)-trans-ACPD. Widely used in studies related to neurotransmissions mediated via group II mGlu receptors.

Biochem/physiol Actions

Primary Target
mGlu₂ & mGlu₃

Warning

Toxicity: Standard Handling (A)

Other Notes

Shinozaki, H., et al. 1989. Br. J. Pharmacol.98, 1213.
Hayashi, Y., et al. 1992. Br. J. Pharmacol.107, 539.
Nakanishi,S. 1992. Science.258, 597.
Watkins, J., et al. 1994. Trends Pharmacol Sci.15, 333.
Schoepp, D.D., et al. 1999. Neuropharmacology.38, 1431.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service