Skip to Content
Merck
All Photos(1)

Key Documents

2116-M

Millipore

Detergent-OUT Med Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NB.24

technique(s)

protein clean-up: suitable

application(s)

sample preparation

General description

The Chemicon Detergent-OUT Kit is a simple high performance method for removing detergents from protein solutions. Removes detergents without significant dilution of protein solution. Simply load the protein solution on the Detergent-OUT Spin column and collect detergent free protein solution. The Med column is suitable for removing detergents from a 1.0 mL solution of 1.5% Triton-X100 detergents (or an equivalent of 15 μL Triton-X100 or 15 mg detergents), provided that both the columns and proteins solutions are prepared in water.

Application

PROTOCOL:

1. Invert the column several times to re-suspend the resin material. Remove the tip of the column and let the liquid drain off.

2. Buffer Equilibration: Detergent-OUT columns are supplied in deionized water. A better result is obtained when the column and protein solution are in water or phosphate buffer. Columns may be equilibrated with other types of buffer. Note: Do not equilibrate the columns with Tris buffer and other such buffers. Apply about 1.0 mL of a desired buffer into the column. Let the buffer drain into collection tube. Repeat this process 3 times.

3. Place the column in a 15mL centrifuge tube. Centrifuge at 1000 xg for 20-30 seconds. Discard the liquid collected in the centrifuge tube.

4. Place the column back in the same centrifuge tube. Carefully apply the protein suspension to the center of the column. Wait for 5 minutes..

5. After loading the column centrifuge at 1000 xg for 20-30 seconds. Collect the liquid comprising purified protein material free from detergent originally present in the sample.

Packaging

10 columns

Components

Detergent-OUT Columns-DTG-100X 10 Med

Storage and Stability

Store at 2-8°C for up to 6 months from date of purchase. To avoid possible microbial contamination, dispense all solutions aseptically.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service