Skip to Content
Merck
All Photos(1)

Documents

12-408

Sigma-Aldrich

Histone H2B Peptide, biotin conjugate, residues 21-41

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.42

Quality Level

manufacturer/tradename

Upstate®

technique(s)

affinity precipitation assay: suitable
immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Application

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes.

Quality

Routinely evaluated as a substrate for histone-modifying enzymes.

Physical form

Lyophilized powder

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The hydroxylation activity of Jmjd6 is required for its homo-oligomerization.
Gang Han,Jiajia Li,Yiqin Wang,Xia Li,Hailei Mao,Yifan Liu,Charlie Degui Chen
Journal of Cellular Biochemistry null
D R Dunbar et al.
Human molecular genetics, 2(10), 1619-1624 (1993-10-01)
Mitochondrial DNA from a 38 year old male with diabetes mellitus and features of mitochondrial dysfunction was analysed and shown to include a population with a partial duplication. The partially duplicated mitochondrial DNA molecules were evident in both muscle and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service