1.13353
Ethyl acetate
Synonym(s):
Ethyl ethanoate, Acetic ester, Ethyl ester
About This Item
Recommended Products
grade
for chromatography
for preparative HPLC
for preparative liquid chromatography
Quality Level
vapor pressure
97 hPa ( 20 °C)
product line
Prepsolv™
Assay
≥99.8% n-heptane basis (GC)
form
liquid
autoignition temp.
460 °C
potency
5620 mg/kg LD50, oral (rat)
>18000 mg/kg LD50, skin (rabbit)
expl. lim.
2.1-11.5 % (v/v)
impurities
≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.05% Water
evapn. residue
≤5.0 mg/L
transmittance
270 nm, ≥50%
300 nm, ≥98%
bp
77 °C/1013 hPa
mp
-83 °C
transition temp
flash point -4 °C
solubility
85.3 g/L
density
0.9 g/cm3 at 20 °C
application(s)
food and beverages
pharmaceutical
format
neat
storage temp.
2-25°C
Related Categories
General description
Preparation Note
Storage and Stability
Analysis Note
Identity (IR):conforms
Evaporation residue:≤ 5.0 mg/l
Water:≤ 0.05 %
Acidity:≤ 0.0002 meq/g
Alkalinity:≤ 0.0002 meq/g
Transmission (at 270 nm):≥ 50 %
Transmission (from 300 nm):≥ 98 %
Filtered by 0.2 μm filter
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
24.8 °F
Flash Point(C)
-4 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service