Skip to Content
Merck
All Photos(2)

Key Documents

1.08326

Supelco

Toluene

dried (max. 0.005% H₂O) SeccoSolv

Synonym(s):

Toluol, Methylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
MDL number:
UNSPSC Code:
12191501
EC Index Number:
203-625-9
Assay:
≥99.9% (GC)
technique(s):
DNA amplification: suitable
application(s):
peptide synthesis
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

technique(s)

DNA amplification: suitable

impurities

≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤0.005% Water
≤0.01% Benzene (GC)
≤0.05% Ethylbenzene (GC)
≤0.1% Xylene (GC)

evapn. residue

≤10 mg/L

color

APHA: ≤10

refractive index

n/D 1.496 (lit.)

kinematic viscosity

0.7 cSt(20 °C)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

transition temp

flash point 4 °C

solubility

0.52 g/L

density

0.865 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Colour: ≤ 10 Hazen
Acidity: ≤ 0.0005 meq/g
Alkalinity: ≤ 0.0002 meq/g
Benzene (GC): ≤ 0.01 %
Ethylbenzene (GC): ≤ 0.05 %
Xylene (GC): ≤ 0.1 %
Evaporation residue: ≤ 10 mg/l
Water: ≤ 0.005 %

Legal Information

SECCOSOLV is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F

Flash Point(C)

4.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service