Skip to Content
Merck
All Photos(1)

Documents

07-1507

Sigma-Aldrich

Anti-phospho-Mypt1 (Ser507) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein phosphatase 1 regulatory subunit 12A, Myosin phosphatase-targeting subunit 1, Myosin phosphatase target subunit 1, Protein phosphatase myosin-binding subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology), chicken (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer507)

Gene Information

human ... PPP1R12A(4659)

General description

Myosin phosphatase targeting subunit 1 (MYPT1) is a regulatory targeting subunit that is thought to be responsible for targeting the holoenzyme to the proper positions in muscle and moving PP1 to myosin. MYPT1 is the major subunit involved in myosin light chain phosphatase (MLCP). It is also essential for vasodilation caused by nitric oxide (NO). A decrease in MYPT1 has been shown to play a role in increased tolerance of NO in pulmonary arteries.

Specificity

Other homologies: Rat (89% sequence homology).
This antibody recognizes Mypt1 phosphorylated at (Ser507).

Immunogen

Epitope: Phosphorylated (Ser507)
KLH-conjugated linear peptide corresponding to human Mypt1 phosphorylated at (Ser507).

Application

Research Category
Signaling

Cell Structure
Research Sub Category
Developmental Signaling

Muscle Physiology
This Anti-phospho-Mypt1 (Ser507) Antibody is validated for use in WB for the detection of phospho-Mypt1 (Ser507).

Quality

Evaluated by Western Blot in untreated and PMA-treated NIH/3T3 cell lysates.

Western Blot Analysis: 2 µg/mL of this antibody detected Mypt1 in 10 µg of untreated and PMA-treated NIH/3T3 cell lysates.

Target description

~120 kDa observed.
Uniprot gives a calculated molecular weight of 115 kDa, which can be observed at higher molecular weights due to hyperphosphorylation. Uncharacterized bands appear at ~30 kDa and ~130 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4) and 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Untreated and PMA-treated NIH/3T3 cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Cao Liu et al.
Clinical and experimental pharmacology & physiology, 44(10), 1053-1059 (2017-07-07)
The effects of hypertonic solution on airway smooth muscle (ASM) contraction and the underlying mechanisms are largely unknown. We found that hypertonic saline (HS) inhibited acetylcholine (ACh)-induced contraction of ASM from the mouse trachea and human bronchi. In single mouse
Yuan-Yuan Chen et al.
Journal of pharmacological sciences, 142(2), 60-68 (2019-12-18)
The purpose of this study was to screen a bronchodilator from old drugs and elucidate the underlying mechanism. Paracetamol (acetaminophen) is a widely used analgesic and antipyretic drug. It has been reported that it inhibits the generation of prostaglandin and
Jun-Ying Qiu et al.
Cell biology international, 44(9), 1870-1880 (2020-05-22)
The objective of this project was to find a bronchodilatory compound from herbs and clarify the mechanism. We found that the ethanol extract of Folium Sennae (EEFS) can relax airway smooth muscle (ASM). EEFS inhibited ASM contraction, induced by acetylcholine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service