Skip to Content
Merck
All Photos(2)

Documents

06-1376

Sigma-Aldrich

Anti-SLBP Antibody

serum, from rabbit

Synonym(s):

Histone stem-loop-binding protein, hairpin binding protein, histone binding protein 2, histone stem-loop binding protein, stem-loop (histone) binding protein, stem-loop binding protein, HBP, Histone RNA hairpin-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

rat

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLBP(7884)

General description

The SLPB gene (stem-loop binding protein) encodes a protein that binds to the stem-loop structure of replication-dependent histone pre-mRNAs. Histone mRNA lack a polyadenylation signal. Consequently, the interaction of this protein with histone pre-mRNA contributes to efficient 3′ end processing by stabilizing the complex between the pre-mRNA and U7 small nuclear ribonucleoprotein (snRNP). Expression of this protein is regulated during the cell cycle, increasing more than 10-fold during the latter part of G1.

Specificity

Antibody recognizes SLBP.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human SLBP.

Application

Immunocytochemistry:
Representative lot data.
Confocal fluorescent analysis of NIH/3T3 cells using 06-1376 (Red). Actin filaments have been labeled with AlexaFluor488 -Phalloidin (Green). Nucleus is stained with DAPI (Blue). This antibody positively stains cytoplasm and nucleus.
Research Sub Category
RNA Metabolism & Binding Proteins

Histones
Use Anti-SLBP Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC to detect SLBP also known as Histone stem-loop-binding protein, HBP, Histone RNA hairpin-binding protein.

Quality

Evaluated by Western Blotting in C2C12 cell lysates.

Western Blotting Analysis: A 1:1000 dilution of this antibody detected SLBP in C2C12 cell lysates.

Target description

38 kDa

Physical form

Serum in buffer containing 0.05% sodium azide

Analysis Note

Control
C2C12 cell lysates.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service