Skip to Content
Merck
All Photos(1)

Key Documents

MAN238929960

Manchester Organics

3-Bromo-6-piperidin-1-yl-imidazo[1,2-b]pyridazine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 158.32
1 G
CHF 261.82
2.5 G
CHF 524.02

Vendor SKU: C19801-500MG

CHF 158.32


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
500 MG
CHF 158.32
1 G
CHF 261.82
2.5 G
CHF 524.02

About This Item

CAS Number:
Molecular Weight:
281.16
MDL number:

Vendor SKU: C19801-500MG

CHF 158.32


Ships in 10 days - from Aldrich Partner.

Iupac Name

3-bromo-6-(1-piperidinyl)imidazo[1,2-b]pyridazine

Inchi Code

1S/C11H13BrN4/c12-9-8-13-10-4-5-11(14-16(9)10)15-6-2-1-3-7-15/h4-5,8H,1-3,6-7H2

InChI key

ZOIHOUTZDWHDBD-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/C19801

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service