Skip to Content
Merck
All Photos(1)

Key Documents

MAN182108509

Manchester Organics

3,4-Difluoro-2-methylbenzonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
CHF 96.22
5 G
CHF 130.72
10 G
CHF 192.82

Vendor SKU: D18192-2.5G

CHF 96.22


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
2.5 G
CHF 96.22
5 G
CHF 130.72
10 G
CHF 192.82

About This Item

CAS Number:
Molecular Weight:
153.13
MDL number:

Vendor SKU: D18192-2.5G

CHF 96.22


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

3,4-difluoro-2-methylbenzonitrile

Inchi Code

1S/C8H5F2N/c1-5-6(4-11)2-3-7(9)8(5)10/h2-3H,1H3

InChI key

LMGHUTIUVKVZNH-UHFFFAOYSA-N

Purity

97%

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/D18192

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service