Skip to Content
Merck
All Photos(1)

Key Documents

LIF509446796

Life Chemicals Inc.

tert-butyl 3-hydrazinylpropanoate hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 526.20
500 MG
CHF 812.20
1 G
CHF 1’099.50
2.5 G
CHF 2’157.70
5 G
CHF 3’217.20
10 G
CHF 4’544.50

Vendor SKU: F8889-5612-250MG

CHF 526.20


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CHF 526.20
500 MG
CHF 812.20
1 G
CHF 1’099.50
2.5 G
CHF 2’157.70
5 G
CHF 3’217.20
10 G
CHF 4’544.50

About This Item

CAS Number:
Molecular Weight:
196.68
MDL number:

Vendor SKU: F8889-5612-250MG

CHF 526.20


Ships in 21 days - from Aldrich Partner.

Iupac Name

tert-butyl 3-hydrazineylpropanoate hydrochloride

Inchi Code

1S/C7H16N2O2.ClH/c1-7(2,3)11-6(10)4-5-9-8;/h9H,4-5,8H2,1-3H3;1H

InChI key

LCBBWUYXNLPZIH-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

solid

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service