Skip to Content
Merck
All Photos(1)

Key Documents

KEY090496858

Key Organics / BIONET

tert-butyl 3-(hydroxymethyl)-1H-indole-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 238.00

Vendor SKU: 11Z-0700-5G

CHF 238.00


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
5 G
CHF 238.00

About This Item

CAS Number:
Molecular Weight:
247.29
MDL number:

Vendor SKU: 11Z-0700-5G

CHF 238.00


Ships in 21 days - from Aldrich Partner.

Iupac Name

tert-butyl 3-(hydroxymethyl)-1H-indole-1-carboxylate

Inchi Code

1S/C14H17NO3/c1-14(2,3)18-13(17)15-8-10(9-16)11-6-4-5-7-12(11)15/h4-8,16H,9H2,1-3H3

InChI key

OOVPQKQFSDFRFA-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Low Melting Solid

Product Link

https://www.keyorganics.net/sku/11Z-0700

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/11Z-0700


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service