Skip to Content
Merck
All Photos(1)

Key Documents

KEY087886581

Key Organics / BIONET

2-bromo-1-[2,3-dichloro-4-(ethanesulfonyl)phenyl]ethan-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 463.30
1 G
CHF 710.80
2 G
CHF 1’370.80
5 G
CHF 3’020.80
10 G
CHF 5’330.80

Vendor SKU: 2M-533S-500MG

CHF 463.30


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
500 MG
CHF 463.30
1 G
CHF 710.80
2 G
CHF 1’370.80
5 G
CHF 3’020.80
10 G
CHF 5’330.80

About This Item

CAS Number:
Molecular Weight:
360.06
MDL number:

Vendor SKU: 2M-533S-500MG

CHF 463.30


Ships in 21 days - from Aldrich Partner.

mp

113 - 115

Iupac Name

2-bromo-1-[2,3-dichloro-4-(ethylsulfonyl)phenyl]ethanone

Inchi Code

1S/C10H9BrCl2O3S/c1-2-17(15,16)8-4-3-6(7(14)5-11)9(12)10(8)13/h3-4H,2,5H2,1H3

InChI key

JWJCTECZTPRUEC-UHFFFAOYSA-N

Purity

90%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/2M-533S

Melting Point

113 - 115

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/2M-533S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service