Skip to Content
Merck
All Photos(1)

Key Documents

KEY061970164

Key Organics / BIONET

2-butyl-1,3-benzoxazole-5-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 208.30
1 G
CHF 298.30
2 G
CHF 545.80

Vendor SKU: 10Y-0833-500MG

CHF 208.30


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
500 MG
CHF 208.30
1 G
CHF 298.30
2 G
CHF 545.80

About This Item

CAS Number:
Molecular Weight:
219.24
MDL number:

Vendor SKU: 10Y-0833-500MG

CHF 208.30


Ships in 21 days - from Aldrich Partner.

mp

136 - 138

Iupac Name

2-butyl-1,3-benzoxazole-5-carboxylic acid

Inchi Code

1S/C12H13NO3/c1-2-3-4-11-13-9-7-8(12(14)15)5-6-10(9)16-11/h5-7H,2-4H2,1H3,(H,14,15)

InChI key

DMLAEJJVBPVLOK-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/10Y-0833

Melting Point

136 - 138

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/10Y-0833


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service