JWPH3249E71F
J & W Pharmlab LLC
2-Benzyl-2,7-diaza-spiro[4.4]nonane dihydrochloride
Select a Size
Vendor SKU: 75R0020S-100MG
CHF 226.41
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
Vendor SKU: 75R0020S-100MG
CHF 226.41
Notify Me
Get notified when this item is ready to ship via email.
Iupac Name
2-benzyl-2,7-diazaspiro[4.4]nonane dihydrochloride
Inchi Code
1S/C14H20N2.2ClH/c1-2-4-13(5-3-1)10-16-9-7-14(12-16)6-8-15-11-14;;/h1-5,15H,6-12H2;2*1H
InChI key
HMCPNAXNVHQMBL-UHFFFAOYSA-N
Purity
Country of Origin
Physical Form
Product Link
MSDS
https://www.jwpharmlab.com/home/product/75R0020S
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service