Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249E71B

J & W Pharmlab LLC

2-Phenyl-2,7-diaza-spiro[4.4]nonane hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 456.71
100 MG
CHF 589.84
250 MG
CHF 856.09
500 MG
CHF 1’191.56
1 G
CHF 1’921.09

Vendor SKU: 75R0019M-50MG

CHF 456.71


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CHF 456.71
100 MG
CHF 589.84
250 MG
CHF 856.09
500 MG
CHF 1’191.56
1 G
CHF 1’921.09

About This Item

CAS Number:
Molecular Weight:
238.76
MDL number:

Vendor SKU: 75R0019M-50MG

CHF 456.71


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-phenyl-2,7-diazaspiro[4.4]nonane hydrochloride

Inchi Code

1S/C13H18N2.ClH/c1-2-4-12(5-3-1)15-9-7-13(11-15)6-8-14-10-13;/h1-5,14H,6-11H2;1H

InChI key

BCKKWKAASJOXMC-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Off White Solid

Product Link

https://www.jwpharmlab.com/home/product/75R0019M

MSDS

https://www.jwpharmlab.com/home/product/75R0019M


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service