Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249DD8D

J & W Pharmlab LLC

7-Nitro-3,4-dihydro-1H-quinoxalin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 323.59
100 MG
CHF 456.71
250 MG
CHF 722.96
500 MG
CHF 989.21
1 G
CHF 1’388.59
5 G
CHF 5’382.34

Vendor SKU: 65R0056-50MG

CHF 323.59


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 MG
CHF 323.59
100 MG
CHF 456.71
250 MG
CHF 722.96
500 MG
CHF 989.21
1 G
CHF 1’388.59
5 G
CHF 5’382.34

About This Item

CAS Number:
Molecular Weight:
193.16
MDL number:

Vendor SKU: 65R0056-50MG

CHF 323.59


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

7-nitro-3,4-dihydro-2(1H)-quinoxalinone

Inchi Code

1S/C8H7N3O3/c12-8-4-9-6-2-1-5(11(13)14)3-7(6)10-8/h1-3,9H,4H2,(H,10,12)

InChI key

MLRCQCJQHUFFBS-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

orange solid

Product Link

https://www.jwpharmlab.com/home/product/65R0056

MSDS

https://www.jwpharmlab.com/home/product/65R0056


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service