Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249D187

J & W Pharmlab LLC

1-Isopropyl-1H-indazole-3-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 267.68
100 MG
CHF 342.23
250 MG
CHF 492.66
500 MG
CHF 790.86
1 G
CHF 1’388.59
5 G
CHF 5’382.34

Vendor SKU: 45R0361-50MG

CHF 267.68


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CHF 267.68
100 MG
CHF 342.23
250 MG
CHF 492.66
500 MG
CHF 790.86
1 G
CHF 1’388.59
5 G
CHF 5’382.34

About This Item

CAS Number:
Molecular Weight:
204.23
MDL number:

Vendor SKU: 45R0361-50MG

CHF 267.68


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-isopropyl-1H-indazole-3-carboxylic acid

Inchi Code

1S/C11H12N2O2/c1-7(2)13-9-6-4-3-5-8(9)10(12-13)11(14)15/h3-7H,1-2H3,(H,14,15)

InChI key

FHUPHXAYPNYFIU-UHFFFAOYSA-N

Purity

98%

Country of Origin

US

Physical Form

White Solid

Product Link

https://www.jwpharmlab.com/home/product/45R0361

MSDS

https://www.jwpharmlab.com/home/product/45R0361


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service