Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249C257

J & W Pharmlab LLC

5,5,5-Trifluoro-3-oxo-pentanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
184.11
MDL number:

Pricing and availability is not currently available.

Iupac Name

methyl 5,5,5-trifluoro-3-oxopentanoate

Inchi Code

1S/C6H7F3O3/c1-12-5(11)2-4(10)3-6(7,8)9/h2-3H2,1H3

InChI key

NIQRIYYEIXZIJI-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Clear Liquid

Product Link

https://www.jwpharmlab.com/home/product/05R0350

MSDS

https://www.jwpharmlab.com/home/product/05R0350


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service