Skip to Content
Merck
All Photos(1)

Key Documents

ENAH961CE507

Enamine

1-[({6-fluorospiro[3.3]heptan-2-yl}methyl)sulfanyl]ethan-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 626.10
100 MG
CHF 804.20

Vendor SKU: EN300-27685909-50MG

CHF 626.10


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CHF 626.10
100 MG
CHF 804.20

About This Item

CAS Number:
Molecular Weight:
202.29
MDL number:

Vendor SKU: EN300-27685909-50MG

CHF 626.10


Ships in 14 days - from Aldrich Partner.

Iupac Name

S-((6-fluorospiro[3.3]heptan-2-yl)methyl) ethanethioate

Inchi Code

1S/C10H15FOS/c1-7(12)13-6-8-2-10(3-8)4-9(11)5-10/h8-9H,2-6H2,1H3

InChI key

MEUIHNOZQJJIRC-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-27685909

MSDS

https://www.enaminestore.com/catalog/EN300-27685909


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service