Skip to Content
Merck
All Photos(1)

Key Documents

ENAH6591F826

Enamine

2-(4-hydroxypiperidin-1-yl)benzoic acid hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 344.00
250 MG
CHF 474.00
500 MG
CHF 719.70
1 G
CHF 909.50
2.5 G
CHF 1’741.50
5 G
CHF 2’556.60
10 G
CHF 3’768.20

Vendor SKU: EN300-7445303-100MG

CHF 344.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 344.00
250 MG
CHF 474.00
500 MG
CHF 719.70
1 G
CHF 909.50
2.5 G
CHF 1’741.50
5 G
CHF 2’556.60
10 G
CHF 3’768.20

About This Item

CAS Number:
Molecular Weight:
257.72
MDL number:

Vendor SKU: EN300-7445303-100MG

CHF 344.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-(4-hydroxypiperidin-1-yl)benzoic acid hydrochloride

Inchi Code

1S/C12H15NO3.ClH/c14-9-5-7-13(8-6-9)11-4-2-1-3-10(11)12(15)16;/h1-4,9,14H,5-8H2,(H,15,16);1H

InChI key

WLUCOKWXKODJMJ-UHFFFAOYSA-N

storage temp.

RT

Salt Data

Cl

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-7445303

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-7445303


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service