Skip to Content
Merck
All Photos(1)

Key Documents

ENAH5802EAE6

Enamine

3-hydrazinyl-2-methoxypyridine dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 68.40
250 MG
CHF 78.80
500 MG
CHF 98.30
1 G
CHF 113.90
2.5 G
CHF 177.60
5 G
CHF 284.20

Vendor SKU: EN300-7438417-100MG

CHF 68.40


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 68.40
250 MG
CHF 78.80
500 MG
CHF 98.30
1 G
CHF 113.90
2.5 G
CHF 177.60
5 G
CHF 284.20

About This Item

CAS Number:
Molecular Weight:
212.08
MDL number:

Vendor SKU: EN300-7438417-100MG

CHF 68.40


Ships in 14 days - from Aldrich Partner.

Iupac Name

3-hydrazineyl-2-methoxypyridine dihydrochloride

Inchi Code

1S/C6H9N3O.2ClH/c1-10-6-5(9-7)3-2-4-8-6;;/h2-4,9H,7H2,1H3;2*1H

InChI key

CXVOTUIYGMGVFJ-UHFFFAOYSA-N

storage temp.

RT

Salt Data

Cl

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-7438417

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-7438417


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service