Skip to Content
Merck
All Photos(1)

Key Documents

ENAH11DD030D

Enamine

(4,5,6,7-tetrahydro-1-benzothiophen-3-yl)methanol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 461.00
250 MG
CHF 640.40
500 MG
CHF 983.60
1 G
CHF 1’247.50
2.5 G
CHF 2’401.90
5 G
CHF 3’531.60
10 G
CHF 5’213.80

Vendor SKU: EN300-1223146-100MG

CHF 461.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 461.00
250 MG
CHF 640.40
500 MG
CHF 983.60
1 G
CHF 1’247.50
2.5 G
CHF 2’401.90
5 G
CHF 3’531.60
10 G
CHF 5’213.80

About This Item

CAS Number:
Molecular Weight:
168.26
MDL number:

Vendor SKU: EN300-1223146-100MG

CHF 461.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

4,5,6,7-tetrahydro-1-benzothien-3-ylmethanol

Inchi Code

1S/C9H12OS/c10-5-7-6-11-9-4-2-1-3-8(7)9/h6,10H,1-5H2

InChI key

WTAPYBKGPRPQHV-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

oil

Product Link

https://www.enaminestore.com/catalog/EN300-1223146

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-1223146


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service