Skip to Content
Merck
All Photos(1)

Key Documents

ENAH049F1E60

Enamine

2-chloro-3-ethylpyrazine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 78.80
250 MG
CHF 94.40
500 MG
CHF 121.70
1 G
CHF 143.80
2.5 G
CHF 237.40
5 G
CHF 393.40
10 G
CHF 705.40

Vendor SKU: EN300-312329-100MG

CHF 78.80


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 78.80
250 MG
CHF 94.40
500 MG
CHF 121.70
1 G
CHF 143.80
2.5 G
CHF 237.40
5 G
CHF 393.40
10 G
CHF 705.40

About This Item

CAS Number:
Molecular Weight:
142.59
MDL number:

Vendor SKU: EN300-312329-100MG

CHF 78.80


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-chloro-3-ethylpyrazine

Inchi Code

1S/C6H7ClN2/c1-2-5-6(7)9-4-3-8-5/h3-4H,2H2,1H3

InChI key

JDBIYZFYKSYXPK-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-312329

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-312329


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service