Skip to Content
Merck
All Photos(1)

Key Documents

ENAH03A01CB8

Enamine

1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-4-phenylpiperidine-4-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 68.40
250 MG
CHF 68.40
500 MG
CHF 74.90
1 G
CHF 82.70
2.5 G
CHF 124.30
5 G
CHF 202.30
10 G
CHF 350.50

Vendor SKU: EN300-624095-100MG

CHF 68.40


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 68.40
250 MG
CHF 68.40
500 MG
CHF 74.90
1 G
CHF 82.70
2.5 G
CHF 124.30
5 G
CHF 202.30
10 G
CHF 350.50

About This Item

CAS Number:
Molecular Weight:
427.5
MDL number:

Vendor SKU: EN300-624095-100MG

CHF 68.40


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-[(9H-fluoren-9-ylmethoxy)carbonyl]-4-phenyl-4-piperidinecarboxylic acid

Inchi Code

1S/C27H25NO4/c29-25(30)27(19-8-2-1-3-9-19)14-16-28(17-15-27)26(31)32-18-24-22-12-6-4-10-20(22)21-11-5-7-13-23(21)24/h1-13,24H,14-18H2,(H,29,30)

InChI key

JWEBOBGECOQGTL-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-624095

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-624095


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service