ENA964491881
Enamine
2-(4-fluorophenoxy)ethane-1-sulfonyl fluoride
Select a Size
Vendor SKU: EN300-727158-100MG
CHF 232.20
Select a Size
About This Item
Vendor SKU: EN300-727158-100MG
CHF 232.20
Iupac Name
2-(4-fluorophenoxy)ethane-1-sulfonyl fluoride
Inchi Code
1S/C8H8F2O3S/c9-7-1-3-8(4-2-7)13-5-6-14(10,11)12/h1-4H,5-6H2
InChI key
DZSXWXZUCOECNR-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
MSDS
https://www.enaminestore.com/catalog/EN300-727158
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service