Skip to Content
Merck
All Photos(1)

Key Documents

ENA580354376

Enamine

1,6-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 185.40
250 MG
CHF 245.20
500 MG
CHF 429.80
1 G
CHF 557.20
2.5 G
CHF 1’047.30
5 G
CHF 1’529.60
10 G
CHF 2’245.90

Vendor SKU: EN300-322001-100MG

CHF 185.40


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 185.40
250 MG
CHF 245.20
500 MG
CHF 429.80
1 G
CHF 557.20
2.5 G
CHF 1’047.30
5 G
CHF 1’529.60
10 G
CHF 2’245.90

About This Item

CAS Number:
Molecular Weight:
165.15
MDL number:

Vendor SKU: EN300-322001-100MG

CHF 185.40


Ships in 14 days - from Aldrich Partner.

mp

297-298

Iupac Name

1,6-dimethyl-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile

Inchi Code

1S/C7H7N3O2/c1-4-5(3-8)6(11)9-7(12)10(4)2/h1-2H3,(H,9,11,12)

InChI key

KKMBCMVGVAEMEF-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-322001

Melting Point

297-298

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-322001


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service