Skip to Content
Merck
All Photos(1)

Key Documents

ENA514506597

Enamine

2-isocyanatooxane

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 432.40
250 MG
CHF 597.50
500 MG
CHF 916.00
1 G
CHF 1’161.70
2.5 G
CHF 2’390.20
5 G
CHF 4’679.50
10 G
CHF 9’090.40

Vendor SKU: EN300-274207-100MG

CHF 432.40


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 432.40
250 MG
CHF 597.50
500 MG
CHF 916.00
1 G
CHF 1’161.70
2.5 G
CHF 2’390.20
5 G
CHF 4’679.50
10 G
CHF 9’090.40

About This Item

CAS Number:
Molecular Weight:
127.14
MDL number:

Vendor SKU: EN300-274207-100MG

CHF 432.40


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-isocyanatotetrahydro-2H-pyran

Inchi Code

1S/C6H9NO2/c8-5-7-6-3-1-2-4-9-6/h6H,1-4H2

InChI key

VXVWIRKAABQIGS-UHFFFAOYSA-N

storage temp.

-10

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-274207

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service