Skip to Content
Merck
All Photos(1)

Key Documents

ENA497375146

Enamine

1-cyclopropyl-3,3,3-trifluoropropan-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 644.30
100 MG
CHF 828.90
250 MG
CHF 1’165.60
500 MG
CHF 1’813.00
1 G
CHF 2’309.60
2.5 G
CHF 4’483.20
5 G
CHF 6’611.30
10 G
CHF 9’783.30

Vendor SKU: EN300-253979-50MG

CHF 644.30


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CHF 644.30
100 MG
CHF 828.90
250 MG
CHF 1’165.60
500 MG
CHF 1’813.00
1 G
CHF 2’309.60
2.5 G
CHF 4’483.20
5 G
CHF 6’611.30
10 G
CHF 9’783.30

About This Item

CAS Number:
Molecular Weight:
152.12
MDL number:

Vendor SKU: EN300-253979-50MG

CHF 644.30


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-cyclopropyl-3,3,3-trifluoro-1-propanone

Inchi Code

1S/C6H7F3O/c7-6(8,9)3-5(10)4-1-2-4/h4H,1-3H2

InChI key

BFZARKPIFOUSRN-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-253979

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service