Skip to Content
Merck
All Photos(1)

Key Documents

ENA490047670

Enamine

tetramethyl-1,4-dioxane-2,6-dione

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 476.60
100 MG
CHF 687.20
250 MG
CHF 964.10
500 MG
CHF 1’493.20
1 G
CHF 1’902.70
2.5 G
CHF 3’686.30
5 G
CHF 5’430.90

Vendor SKU: EN300-249032-50MG

CHF 476.60


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CHF 476.60
100 MG
CHF 687.20
250 MG
CHF 964.10
500 MG
CHF 1’493.20
1 G
CHF 1’902.70
2.5 G
CHF 3’686.30
5 G
CHF 5’430.90

About This Item

CAS Number:
Molecular Weight:
172.18
MDL number:

Vendor SKU: EN300-249032-50MG

CHF 476.60


Ships in 14 days - from Aldrich Partner.

mp

71-72

Iupac Name

3,3,5,5-tetramethyl-1,4-dioxane-2,6-dione

Inchi Code

1S/C8H12O4/c1-7(2)5(9)11-6(10)8(3,4)12-7/h1-4H3

InChI key

CGVYQNFYKAABLQ-UHFFFAOYSA-N

storage temp.

4

Purity

80

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-249032

Melting Point

71-72

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-249032


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service