Skip to Content
Merck
All Photos(1)

Key Documents

ENA444725308

Enamine

3-bromo-2,2-difluoropropan-1-ol

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 774.30
100 MG
CHF 1’000.50
250 MG
CHF 1’404.80
500 MG
CHF 2’190.00
1 G
CHF 2’794.50
2.5 G
CHF 5’436.10
5 G
CHF 8’019.20
10 G
CHF 11’869.80

Vendor SKU: EN300-203452-50MG

CHF 774.30


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CHF 774.30
100 MG
CHF 1’000.50
250 MG
CHF 1’404.80
500 MG
CHF 2’190.00
1 G
CHF 2’794.50
2.5 G
CHF 5’436.10
5 G
CHF 8’019.20
10 G
CHF 11’869.80

About This Item

CAS Number:
Molecular Weight:
174.97
MDL number:

Vendor SKU: EN300-203452-50MG

CHF 774.30


Ships in 14 days - from Aldrich Partner.

Iupac Name

3-bromo-2,2-difluoro-1-propanol

Inchi Code

1S/C3H5BrF2O/c4-1-3(5,6)2-7/h7H,1-2H2

InChI key

JCDHRVTXINIMAY-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-203452

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-203452


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service