Skip to Content
Merck
All Photos(1)

Key Documents

ENA428073745

Enamine

4-(aminomethyl)pyridine-2-carbonitrile dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 357.00
250 MG
CHF 489.60
500 MG
CHF 745.70
1 G
CHF 943.30
2.5 G
CHF 1’953.40
5 G
CHF 3’834.50
10 G
CHF 7’460.20

Vendor SKU: EN300-182515-100MG

CHF 357.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 357.00
250 MG
CHF 489.60
500 MG
CHF 745.70
1 G
CHF 943.30
2.5 G
CHF 1’953.40
5 G
CHF 3’834.50
10 G
CHF 7’460.20

About This Item

CAS Number:
Molecular Weight:
206.07
MDL number:

Vendor SKU: EN300-182515-100MG

CHF 357.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

4-(aminomethyl)picolinonitrile dihydrochloride

Inchi Code

1S/C7H7N3.2ClH/c8-4-6-1-2-10-7(3-6)5-9;;/h1-3H,4,8H2;2*1H

InChI key

LHQAMQIAECCSMV-UHFFFAOYSA-N

storage temp.

RT

Salt Data

Cl

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-182515

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-182515


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service