Skip to Content
Merck
All Photos(1)

Key Documents

ENA428072872

Enamine

1-propyl-1,4-diazepan-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 126.90
250 MG
CHF 159.40
500 MG
CHF 260.80
1 G
CHF 362.20
2.5 G
CHF 666.40
5 G
CHF 964.10

Vendor SKU: EN300-154166-100MG

CHF 126.90


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 126.90
250 MG
CHF 159.40
500 MG
CHF 260.80
1 G
CHF 362.20
2.5 G
CHF 666.40
5 G
CHF 964.10

About This Item

CAS Number:
Molecular Weight:
156.23
MDL number:

Vendor SKU: EN300-154166-100MG

CHF 126.90


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-propyl-1,4-diazepan-2-one

Inchi Code

1S/C8H16N2O/c1-2-5-10-6-3-4-9-7-8(10)11/h9H,2-7H2,1H3

InChI key

ONMRUIGEYUMICY-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-154166

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-154166


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service