ENA410543423
Enamine
2,3,5-trichloro-6-(trichloromethyl)pyridin-4-amine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
118-120
Iupac Name
2,3,5-trichloro-6-(trichloromethyl)-4-pyridinamine
Inchi Code
1S/C6H2Cl6N2/c7-1-3(13)2(8)5(9)14-4(1)6(10,11)12/h(H2,13,14)
InChI key
VNJVZJNBTWPKNX-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-16703
Melting Point
118-120
MSDS
https://www.enaminestore.com/catalog/EN300-16703
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service