Skip to Content
Merck
All Photos(1)

Key Documents

ENA405194212

Enamine

bis(N-(propan-2-yl)guanidine); sulfuric acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 347.90
250 MG
CHF 477.90
500 MG
CHF 726.20
1 G
CHF 918.60
2.5 G
CHF 1’088.90
5 G
CHF 1’371.00
10 G
CHF 1’936.50

Vendor SKU: EN300-133295-100MG

CHF 347.90


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 347.90
250 MG
CHF 477.90
500 MG
CHF 726.20
1 G
CHF 918.60
2.5 G
CHF 1’088.90
5 G
CHF 1’371.00
10 G
CHF 1’936.50

About This Item

CAS Number:
Molecular Weight:
300.38
MDL number:

Vendor SKU: EN300-133295-100MG

CHF 347.90


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-isopropylguanidine hemisulfate

Inchi Code

1S/2C4H11N3.H2O4S/c2*1-3(2)7-4(5)6;1-5(2,3)4/h2*3H,1-2H3,(H4,5,6,7);(H2,1,2,3,4)

InChI key

MFIQZHWDRVKGAK-UHFFFAOYSA-N

storage temp.

RT

Salt Data

OS(=O)(=O)O

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-133295

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-133295


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service