Skip to Content
Merck
All Photos(1)

Key Documents

ENA388700180

Enamine

2-[2-(propan-2-yl)pyrimidin-5-yl]ethan-1-amine

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 384.30
100 MG
CHF 548.10
250 MG
CHF 761.30

Vendor SKU: EN300-123833-50MG

CHF 384.30


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CHF 384.30
100 MG
CHF 548.10
250 MG
CHF 761.30

About This Item

CAS Number:
Molecular Weight:
165.24
MDL number:

Vendor SKU: EN300-123833-50MG

CHF 384.30


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-(2-isopropyl-5-pyrimidinyl)ethylamine

Inchi Code

1S/C9H15N3/c1-7(2)9-11-5-8(3-4-10)6-12-9/h5-7H,3-4,10H2,1-2H3

InChI key

GKJVOJSTXXAMMM-UHFFFAOYSA-N

storage temp.

4

Purity

92

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-123833

Pictograms

CorrosionExclamation mark

Signal Word

Danger

MSDS

https://www.enaminestore.com/catalog/EN300-123833


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service