Skip to Content
Merck
All Photos(1)

Key Documents

ENA341291168

Enamine

4-(4-methoxyphenyl)benzonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 94.40
250 MG
CHF 115.20
500 MG
CHF 156.80
1 G
CHF 188.00
2.5 G
CHF 289.40
5 G
CHF 459.70
10 G
CHF 762.60

Vendor SKU: EN300-86160-100MG

CHF 94.40


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 94.40
250 MG
CHF 115.20
500 MG
CHF 156.80
1 G
CHF 188.00
2.5 G
CHF 289.40
5 G
CHF 459.70
10 G
CHF 762.60

About This Item

CAS Number:
Molecular Weight:
209.25
MDL number:

Vendor SKU: EN300-86160-100MG

CHF 94.40


Ships in 14 days - from Aldrich Partner.

mp

101-103

Iupac Name

4'-methoxy[1,1'-biphenyl]-4-carbonitrile

Inchi Code

1S/C14H11NO/c1-16-14-8-6-13(7-9-14)12-4-2-11(10-15)3-5-12/h2-9H,1H3

InChI key

RINYLKSNGRLNCU-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-86160

Melting Point

101-103

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-86160


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service