Skip to Content
Merck
All Photos(1)

Key Documents

ENA295068236

Enamine

N,N-dimethyl-1-[(5-oxo-5,6,7,8-tetrahydronaphthalen-2-yl)sulfanyl]formamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 165.90
250 MG
CHF 220.50
500 MG
CHF 379.10
1 G
CHF 503.90
2.5 G
CHF 944.60
5 G
CHF 1’374.90
10 G
CHF 2’017.10

Vendor SKU: EN300-64467-100MG

CHF 165.90


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 165.90
250 MG
CHF 220.50
500 MG
CHF 379.10
1 G
CHF 503.90
2.5 G
CHF 944.60
5 G
CHF 1’374.90
10 G
CHF 2’017.10

About This Item

CAS Number:
Molecular Weight:
249.33
MDL number:

Vendor SKU: EN300-64467-100MG

CHF 165.90


Ships in 14 days - from Aldrich Partner.

mp

95-97

Iupac Name

S-(5-oxo-5,6,7,8-tetrahydro-2-naphthalenyl) dimethylthiocarbamate

Inchi Code

1S/C13H15NO2S/c1-14(2)13(16)17-10-6-7-11-9(8-10)4-3-5-12(11)15/h6-8H,3-5H2,1-2H3

InChI key

AMZNKLYLFKFXDT-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-64467

Melting Point

95-97

MSDS

https://www.enaminestore.com/catalog/EN300-64467


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service