Skip to Content
Merck
All Photos(1)

Key Documents

ENA238711886

Enamine

2-(1,3,4-thiadiazol-2-ylsulfanyl)acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 146.40
250 MG
CHF 188.00
500 MG
CHF 315.40
1 G
CHF 432.40
2.5 G
CHF 804.20
5 G
CHF 1’169.50
10 G
CHF 1’712.90

Vendor SKU: EN300-49810-100MG

CHF 146.40


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 146.40
250 MG
CHF 188.00
500 MG
CHF 315.40
1 G
CHF 432.40
2.5 G
CHF 804.20
5 G
CHF 1’169.50
10 G
CHF 1’712.90

About This Item

CAS Number:
Molecular Weight:
176.22
MDL number:

Vendor SKU: EN300-49810-100MG

CHF 146.40


Ships in 14 days - from Aldrich Partner.

mp

156-158

Iupac Name

(1,3,4-thiadiazol-2-ylsulfanyl)acetic acid

Inchi Code

1S/C4H4N2O2S2/c7-3(8)1-9-4-6-5-2-10-4/h2H,1H2,(H,7,8)

InChI key

DZXGDEPWZLFUEA-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-49810

Melting Point

156-158

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-49810


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service