Skip to Content
Merck
All Photos(1)

Key Documents

ENA238710412

Enamine

{imidazo[2,1-b][1,3]thiazol-6-yl}methanamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 112.60
250 MG
CHF 141.20
500 MG
CHF 197.10
1 G
CHF 240.00
2.5 G
CHF 444.10
5 G
CHF 783.40
10 G
CHF 1’463.30

Vendor SKU: EN300-34475-100MG

CHF 112.60


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 112.60
250 MG
CHF 141.20
500 MG
CHF 197.10
1 G
CHF 240.00
2.5 G
CHF 444.10
5 G
CHF 783.40
10 G
CHF 1’463.30

About This Item

CAS Number:
Molecular Weight:
153.21
MDL number:

Vendor SKU: EN300-34475-100MG

CHF 112.60


Ships in 14 days - from Aldrich Partner.

mp

32-34

Iupac Name

imidazo[2,1-b][1,3]thiazol-6-ylmethanamine

Inchi Code

1S/C6H7N3S/c7-3-5-4-9-1-2-10-6(9)8-5/h1-2,4H,3,7H2

InChI key

DHSVDAKPQOFXKO-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

oil

Product Link

https://www.enaminestore.com/catalog/EN300-34475

Melting Point

32-34

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-34475


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service