Skip to Content
Merck
All Photos(1)

Key Documents

ENA196830210

Enamine

6-chloro-N-(3-methylbutyl)pyridine-3-sulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 277.70
250 MG
CHF 379.10
500 MG
CHF 623.50
1 G
CHF 787.30
2.5 G
CHF 1’498.40
5 G
CHF 2’195.20

Vendor SKU: EN300-44901-100MG

CHF 277.70


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 277.70
250 MG
CHF 379.10
500 MG
CHF 623.50
1 G
CHF 787.30
2.5 G
CHF 1’498.40
5 G
CHF 2’195.20

About This Item

CAS Number:
Molecular Weight:
262.76
MDL number:

Vendor SKU: EN300-44901-100MG

CHF 277.70


Ships in 14 days - from Aldrich Partner.

Iupac Name

6-chloro-N-isopentyl-3-pyridinesulfonamide

Inchi Code

1S/C10H15ClN2O2S/c1-8(2)5-6-13-16(14,15)9-3-4-10(11)12-7-9/h3-4,7-8,13H,5-6H2,1-2H3

InChI key

IIRPHAKQDZKKFY-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-44901

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-44901


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service