COMH04236681
Combi-Blocks, Inc.
1,3-Dihydroxy-12h-benzo[b]xanthen-12-one
Select a Size
Vendor SKU: QY-7384-1G
CHF 336.90
Select a Size
About This Item
Vendor SKU: QY-7384-1G
CHF 336.90
Iupac Name
1,3-dihydroxy-12H-benzo[b]xanthen-12-one
Inchi Code
1S/C17H10O4/c18-11-7-13(19)16-15(8-11)21-14-6-10-4-2-1-3-9(10)5-12(14)17(16)20/h1-8,18-19H
InChI key
MSAPVEXKCXYFIM-UHFFFAOYSA-N
Purity
Country of Origin
Product Link
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?QY-7384
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service