Skip to Content
Merck
All Photos(1)

Key Documents

COM448622964

Combi-Blocks, Inc.

3-Methoxypropyl Z-L-Alaninamide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 267.68
5 G
CHF 890.70

Vendor SKU: LD-0908-1G

CHF 267.68


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 267.68
5 G
CHF 890.70

About This Item

CAS Number:
Molecular Weight:
294.35
MDL number:

Vendor SKU: LD-0908-1G

CHF 267.68


Ships in 14 days - from Aldrich Partner.

Iupac Name

benzyl (S)-(1-((3-methoxypropyl)amino)-1-oxopropan-2-yl)carbamate

Inchi Code

1S/C15H22N2O4/c1-12(14(18)16-9-6-10-20-2)17-15(19)21-11-13-7-4-3-5-8-13/h3-5,7-8,12H,6,9-11H2,1-2H3,(H,16,18)(H,17,19)/t12-/m0/s1

InChI key

SAVSELWCSLOACS-LBPRGKRZSA-N

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?LD-0908

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?LD-0908


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service